Assay Details
| Gene Symbol: BGAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339590.1 | NP_001325739.1 | 12 - 13 | 1619 | 89 | |
| NM_115144.4 | NP_190852.2 | 12 - 13 | 1581 | 89 | ||
| GenBank mRNA | AF367327.1 | - | 12 - 13 | 1513 | 89 | |
| AJ270298.1 | - | 12 - 13 | 1470 | 89 | ||
| BT000511.1 | - | 12 - 13 | 1424 | 89 | ||
More Information
Set Membership: |
|

BGAL2
Genomic Map