Assay Details
| Gene Symbol: BGAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339590.1 | NP_001325739.1 | 13 - 14 | 1729 | 127 | |
| NM_115144.4 | NP_190852.2 | 13 - 14 | 1691 | 127 | ||
| GenBank mRNA | AF367327.1 | - | 13 - 14 | 1623 | 127 | |
| AJ270298.1 | - | 13 - 14 | 1580 | 127 | ||
| BT000511.1 | - | 13 - 14 | 1534 | 127 | ||
More Information
Set Membership: |
|

BGAL2
Genomic Map