Assay Details
| Gene Symbol: BGAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339590.1 | NP_001325739.1 | 16 - 17 | 2061 | 68 | |
| NM_115144.4 | NP_190852.2 | 16 - 17 | 2023 | 68 | ||
| GenBank mRNA | AF367327.1 | - | 16 - 17 | 1955 | 68 | |
| AJ270298.1 | - | 16 - 17 | 1912 | 68 | ||
| BT000511.1 | - | 16 - 17 | 1866 | 68 | ||
More Information
Set Membership: |
|

BGAL2
Genomic Map