Assay Details
| Gene Symbol: FBA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115153.4 | NP_190861.1 | 2 - 3 | 591 | 86 | |
| GenBank mRNA | AK317380.1 | - | 2 - 3 | 329 | 86 | |
| AY052709.1 | - | 2 - 3 | 329 | 86 | ||
| AY057656.1 | - | 2 - 3 | 326 | 86 | ||
| AY063718.1 | - | 2 - 3 | 261 | 86 | ||
More Information
Set Membership: |
|

FBA8
Genomic Map