Assay Details
| Gene Symbol: SRPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115163.5 | NP_566977.1 | 2 - 3 | 1347 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SRPK4| Gene Symbol: SRPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115163.5 | NP_566977.1 | 2 - 3 | 1347 | 69 | |
Set Membership: |
|