Assay Details
| Gene Symbol: AT3G53190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115179.5 | NP_566979.1 | 5 - 6 | 1421 | 74 | |
| GenBank mRNA | AK221575.1 | - | 5 - 6 | 1292 | 74 | |
More Information
Set Membership: |
|

AT3G53190
Genomic Map