Assay Details
| Gene Symbol: MYB27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339599.1 | NP_001325713.1 | 1 - 2 | 458 | 70 | |
| NM_115180.3 | NP_566980.1 | 2 - 3 | 370 | 70 | ||
| GenBank mRNA | AK117256.1 | - | 2 - 3 | 325 | 70 | |
| AY519599.1 | - | 2 - 3 | 257 | 70 | ||
| BT005257.1 | - | 2 - 3 | 257 | 70 | ||
More Information
Set Membership: |
|

MYB27
Genomic Map