Assay Details
| Gene Symbol: AtCDC48B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115183.4 | NP_190891.1 | 3 - 4 | 484 | 120 | |
| GenBank mRNA | AK228801.1 | - | 3 - 4 | 459 | 120 | |
More Information
Set Membership: |
|

AtCDC48B
Genomic Map