Assay Details
| Gene Symbol: AT3G53580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115218.4 | NP_190926.1 | 3 - 4 | 666 | 65 | |
| GenBank mRNA | AY126996.1 | - | 3 - 4 | 539 | 65 | |
| AY143871.1 | - | 3 - 4 | 513 | 65 | ||
More Information
Set Membership: |
|

AT3G53580
Genomic Map