Assay Details
| Gene Symbol: AT3G53580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115218.4 | NP_190926.1 | 5 - 6 | 900 | 93 | |
| GenBank mRNA | AY126996.1 | - | 5 - 6 | 773 | 93 | |
| AY143871.1 | - | 5 - 6 | 747 | 93 | ||
More Information
Set Membership: |
|

AT3G53580
Genomic Map