Assay Details
| Gene Symbol: AT3G53580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115218.4 | NP_190926.1 | 8 - 9 | 1124 | 59 | |
| GenBank mRNA | AY126996.1 | - | 8 - 9 | 997 | 59 | |
| AY143871.1 | - | 8 - 9 | 971 | 59 | ||
More Information
Set Membership: |
|

AT3G53580
Genomic Map