Assay Details
| Gene Symbol: GCP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339630.1 | NP_001325627.1 | 13 - 14 | 2061 | 102 | |
| NM_001339631.1 | NP_001325626.1 | 12 - 13 | 1975 | 102 | ||
| NM_115236.4 | NP_190944.2 | 13 - 14 | 2150 | 102 | ||
| GenBank mRNA | BT012227.1 | - | 13 - 14 | 2000 | 102 | |
| BT012412.1 | - | 13 - 14 | 1902 | 102 | ||
More Information
Set Membership: |
|

GCP4
Genomic Map