Assay Details
| Gene Symbol: GCP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339630.1 | NP_001325627.1 | 1 - 2 | 330 | 94 | |
| NM_001339631.1 | NP_001325626.1 | 1 - 2 | 328 | 94 | ||
| NM_115236.4 | NP_190944.2 | 1 - 2 | 419 | 94 | ||
| GenBank mRNA | BT012227.1 | - | 1 - 2 | 269 | 94 | |
| BT012412.1 | - | 1 - 2 | 171 | 94 | ||
More Information
Set Membership: |
|

GCP4
Genomic Map