Assay Details
| Gene Symbol: GCP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339630.1 | NP_001325627.1 | 8 - 9 | 1416 | 107 | |
| NM_115236.4 | NP_190944.2 | 8 - 9 | 1505 | 107 | ||
| GenBank mRNA | BT012227.1 | - | 8 - 9 | 1355 | 107 | |
| BT012412.1 | - | 8 - 9 | 1257 | 107 | ||
More Information
Set Membership: |
|

GCP4
Genomic Map