Assay Details
| Gene Symbol: Fes1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115240.5 | NP_190948.1 | 1 - 2 | 230 | 69 | |
| GenBank mRNA | AY087709.1 | - | 1 - 2 | 140 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fes1B| Gene Symbol: Fes1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115240.5 | NP_190948.1 | 1 - 2 | 230 | 69 | |
| GenBank mRNA | AY087709.1 | - | 1 - 2 | 140 | 69 | |
Set Membership: |
|