Assay Details
| Gene Symbol: Fes1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339634.1 | NP_001326231.1 | 4 - 5 | 1072 | 123 | |
| NM_115240.5 | NP_190948.1 | 5 - 6 | 746 | 123 | ||
| GenBank mRNA | AY087709.1 | - | 5 - 6 | 656 | 123 | |
More Information
Set Membership: |
|

Fes1B
Genomic Map