Assay Details
| Gene Symbol: AT3G53850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339639.1 | NP_001327256.1 | 3 - 4 | 501 | 124 | |
| NM_001339640.1 | NP_001327257.1 | 1 - 2 | 249 | 124 | ||
| NM_115245.3 | NP_566990.1 | 2 - 3 | 593 | 124 | ||
| GenBank mRNA | AF412058.1 | - | 2 - 3 | 561 | 124 | |
| AY120697.1 | - | 2 - 3 | 267 | 124 | ||
More Information
Set Membership: |
|

AT3G53850
Genomic Map