Assay Details
| Gene Symbol: PTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339651.1 | NP_001326764.1 | 2 - 3 | 362 | 87 | |
| NM_115274.3 | NP_190982.1 | 3 - 4 | 628 | 87 | ||
| GenBank mRNA | AF360155.1 | - | 3 - 4 | 455 | 87 | |
| AY084773.1 | - | 3 - 4 | 444 | 87 | ||
| AY113884.1 | - | 2 - 3 | 326 | 87 | ||
More Information
Set Membership: |
|

PTR1
Genomic Map