Assay Details
| Gene Symbol: PTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339651.1 | NP_001326764.1 | 3 - 4 | 904 | 63 | |
| NM_115274.3 | NP_190982.1 | 4 - 5 | 1170 | 63 | ||
| GenBank mRNA | AF360155.1 | - | 4 - 5 | 997 | 63 | |
| AY084773.1 | - | 4 - 5 | 986 | 63 | ||
| AY113884.1 | - | 3 - 4 | 868 | 63 | ||
More Information
Set Membership: |
|

PTR1
Genomic Map