Assay Details
| Gene Symbol: SUA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203155.1 | NP_001190084.1 | 16 - 17 | 3185 | 70 | |
| NM_001339653.1 | NP_001325865.1 | 13 - 14 | 3627 | 70 | ||
| NM_001339654.1 | NP_001325863.1 | 14 - 15 | 3537 | 70 | ||
| NM_001339655.1 | NP_001325864.1 | 15 - 16 | 3349 | 70 | ||
| NM_115283.3 | NP_190991.2 | 16 - 17 | 3252 | 70 | ||
| GenBank mRNA | AY070066.1 | - | 16 - 17 | 3180 | 70 | |
| AY096433.1 | - | 14 - 15 | 2836 | 70 | ||
More Information
Set Membership: |
|

SUA
Genomic Map