Assay Details
| Gene Symbol: RGD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203156.1 | NP_001190085.1 | 29 - 30 | 6210 | 94 | |
| NM_115288.8 | NP_190996.3 | 28 - 29 | 6241 | 94 | ||
| GenBank mRNA | AB440793.1 | - | 27 - 28 | 5979 | 94 | |
More Information
Set Membership: |
|

RGD3
Genomic Map