Assay Details
| Gene Symbol: AP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115294.6 | NP_191002.1 | 4 - 5 | 643 | 84 | |
| GenBank mRNA | AY070397.1 | - | 4 - 5 | 446 | 84 | |
| AY142590.1 | - | 4 - 5 | 416 | 84 | ||
| M86357.1 | - | 4 - 5 | 474 | 84 | ||
More Information
Set Membership: |
|

AP3
Genomic Map