Assay Details
| Gene Symbol: SKIP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339667.1 | NP_001326219.1 | 3 - 4 | 582 | 87 | |
| NM_001339668.1 | NP_001326223.1 | 4 - 5 | 519 | 87 | ||
| NM_001339669.1 | NP_001326222.1 | 4 - 5 | 519 | 87 | ||
| NM_115304.3 | NP_566999.1 | 3 - 4 | 582 | 87 | ||
| GenBank mRNA | AF325053.1 | - | 3 - 4 | 481 | 87 | |
| AF347971.1 | - | 3 - 4 | 465 | 87 | ||
| AF347972.1 | - | 3 - 4 | 455 | 87 | ||
| AK227454.1 | - | 3 - 4 | 408 | 87 | ||
| AY085042.1 | - | 3 - 4 | 481 | 87 | ||
| BT005863.1 | - | 3 - 4 | 390 | 87 | ||
More Information
Set Membership: |
|

SKIP5
Genomic Map