Assay Details
| Gene Symbol: GR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115323.4 | NP_191026.1 | 10 - 11 | 1857 | 121 | |
| GenBank mRNA | D14049.1 | - | 10 - 11 | 1763 | 120 | |
More Information
Set Membership: |
|

GR
Genomic Map