Assay Details
| Gene Symbol: TTN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339686.1 | NP_001319748.1 | 17 - 18 | 3921 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTN8| Gene Symbol: TTN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339686.1 | NP_001319748.1 | 17 - 18 | 3921 | 84 | |
Set Membership: |
|