Assay Details
| Gene Symbol: SETH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115326.5 | NP_191029.1 | 2 - 3 | 977 | 90 | |
| GenBank mRNA | AK227009.1 | - | 2 - 3 | 938 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SETH3| Gene Symbol: SETH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115326.5 | NP_191029.1 | 2 - 3 | 977 | 90 | |
| GenBank mRNA | AK227009.1 | - | 2 - 3 | 938 | 90 | |
Set Membership: |
|