Assay Details
| Gene Symbol: MRH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339702.1 | NP_001326426.1 | 1 - 2 | 432 | 103 | |
| NM_001339703.1 | NP_001326427.1 | 1 - 2 | 394 | 103 | ||
| NM_115344.4 | NP_191047.3 | 1 - 2 | 306 | 103 | ||
| GenBank mRNA | AB290928.1 | - | 1 - 2 | 321 | 103 | |
| EU257694.1 | - | 1 - 2 | 306 | 103 | ||
More Information
Set Membership: |
|

MRH2
Genomic Map