Assay Details
| Gene Symbol: PDIL1-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125370.1 | NP_001118842.1 | 1 - 2 | 591 | 68 | |
| NM_115353.5 | NP_191056.2 | 1 - 2 | 625 | 68 | ||
| GenBank mRNA | AJ271376.1 | - | 1 - 2 | 649 | 68 | |
| AK318844.1 | - | 1 - 2 | 593 | 68 | ||
| AY093115.1 | - | 1 - 2 | 591 | 68 | ||
| BT000172.1 | - | 1 - 2 | 573 | 68 | ||
More Information
Set Membership: |
|

PDIL1-3
Genomic Map