Assay Details
| Gene Symbol: PDIL1-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125370.1 | NP_001118842.1 | 4 - 5 | 1004 | 96 | |
| NM_115353.5 | NP_191056.2 | 4 - 5 | 1038 | 96 | ||
| GenBank mRNA | AJ271376.1 | - | 4 - 5 | 1062 | 96 | |
| AK318844.1 | - | 4 - 5 | 1006 | 96 | ||
| AY093115.1 | - | 4 - 5 | 1004 | 96 | ||
| BT000172.1 | - | 4 - 5 | 986 | 96 | ||
More Information
Set Membership: |
|

PDIL1-3
Genomic Map