Assay Details
| Gene Symbol: TT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115370.4 | NP_191072.1 | 3 - 4 | 703 | 80 | |
| GenBank mRNA | AJ418046.1 | - | 3 - 4 | 572 | 80 | |
| BT004265.1 | - | 3 - 4 | 572 | 80 | ||
| BT005528.1 | - | 3 - 4 | 521 | 80 | ||
More Information
Set Membership: |
|

TT5
Genomic Map