Assay Details
| Gene Symbol: RPL23AB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084824.1 | NP_001078293.1 | 3 - 4 | 436 | 123 | |
| NM_001084825.1 | NP_001078294.1 | 3 - 4 | 481 | 123 | ||
| NM_115386.3 | NP_191088.1 | 3 - 4 | 600 | 123 | ||
| GenBank mRNA | AY050445.1 | - | 3 - 4 | 432 | 123 | |
| AY085736.1 | - | 3 - 4 | 498 | 123 | ||
| BT000478.1 | - | 2 - 3 | 381 | 123 | ||
More Information
Set Membership: |
|

RPL23AB
Genomic Map