Assay Details
| Gene Symbol: PBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203168.1 | NP_001190097.1 | 4 - 5 | 760 | 70 | |
| NM_115403.4 | NP_191105.2 | 4 - 5 | 1107 | 70 | ||
More Information
Set Membership: |
|

PBL1
Genomic Map