Assay Details
| Gene Symbol: SCL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115404.6 | NP_567021.1 | 2 - 3 | 276 | 69 | |
| GenBank mRNA | AY086131.1 | - | 2 - 3 | 254 | 69 | |
| AY099649.1 | - | 2 - 3 | 223 | 69 | ||
| AY128836.1 | - | 1 - 2 | 172 | 69 | ||
More Information
Set Membership: |
|

SCL30
Genomic Map