Assay Details
| Gene Symbol: SCL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115404.6 | NP_567021.1 | 6 - 7 | 641 | 70 | |
| GenBank mRNA | AY086131.1 | - | 6 - 7 | 619 | 70 | |
| AY099649.1 | - | 6 - 7 | 588 | 70 | ||
| AY128836.1 | - | 5 - 6 | 537 | 70 | ||
More Information
Set Membership: |
|

SCL30
Genomic Map