Assay Details
| Gene Symbol: RBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115408.4 | NP_567023.1 | 3 - 4 | 586 | 117 | |
| GenBank mRNA | AK226780.1 | - | 3 - 4 | 530 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBL| Gene Symbol: RBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115408.4 | NP_567023.1 | 3 - 4 | 586 | 117 | |
| GenBank mRNA | AK226780.1 | - | 3 - 4 | 530 | 117 | |
Set Membership: |
|