Assay Details
| Gene Symbol: RBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339730.1 | NP_001326972.1 | 4 - 5 | 1513 | 154 | |
| NM_115408.4 | NP_567023.1 | 5 - 6 | 1377 | 154 | ||
| GenBank mRNA | AK226780.1 | - | 5 - 6 | 1321 | 154 | |
More Information
Set Membership: |
|

RBL
Genomic Map