Assay Details
| Gene Symbol: RBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339730.1 | NP_001326972.1 | 6 - 7 | 2041 | 74 | |
| NM_115408.4 | NP_567023.1 | 7 - 8 | 1905 | 74 | ||
| GenBank mRNA | AK226780.1 | - | 7 - 8 | 1849 | 74 | |
More Information
Set Membership: |
|

RBL
Genomic Map