Assay Details
| Gene Symbol: P5CS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125374.1 | NP_001118846.1 | 15 - 16 | 1816 | 110 | |
| NM_115419.5 | NP_191120.2 | 15 - 16 | 2018 | 110 | ||
| GenBank mRNA | AY091766.1 | - | 15 - 16 | 1816 | 110 | |
| Y09355.1 | - | 1786 | 110 | |||
More Information
Set Membership: |
|

P5CS2
Genomic Map