Assay Details
| Gene Symbol: P5CS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115419.5 | NP_191120.2 | 16 - 17 | 2176 | 101 | |
| GenBank mRNA | AY091766.1 | - | 16 - 17 | 1974 | 101 | |
| Y09355.1 | - | 1944 | 101 | |||
More Information
Set Membership: |
|

P5CS2
Genomic Map