Assay Details
| Gene Symbol: LAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339748.1 | NP_001319763.1 | 9 - 10 | 1742 | 60 | |
| NM_001339749.1 | NP_001326016.1 | 8 - 9 | 1931 | 60 | ||
| NM_001339750.1 | NP_001326018.1 | 8 - 9 | 1931 | 60 | ||
| NM_001339751.1 | NP_001326017.1 | 10 - 11 | 1220 | 60 | ||
| NM_115443.4 | NP_567027.2 | 10 - 11 | 1226 | 60 | ||
| NM_202716.3 | NP_974445.2 | 11 - 12 | 1439 | 60 | ||
More Information
Set Membership: |
|

LAF3
Genomic Map