Assay Details
| Gene Symbol: LAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339748.1 | NP_001319763.1 | 12 - 13 | 2045 | 62 | |
| NM_001339750.1 | NP_001326018.1 | 11 - 12 | 2234 | 62 | ||
| NM_001339751.1 | NP_001326017.1 | 13 - 14 | 1523 | 62 | ||
| NM_115443.4 | NP_567027.2 | 13 - 14 | 1529 | 62 | ||
| NM_202716.3 | NP_974445.2 | 14 - 15 | 1742 | 62 | ||
More Information
Set Membership: |
|

LAF3
Genomic Map