Assay Details
| Gene Symbol: LAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339748.1 | NP_001319763.1 | 2 - 3 | 643 | 77 | |
| NM_001339749.1 | NP_001326016.1 | 2 - 3 | 643 | 77 | ||
| NM_001339750.1 | NP_001326018.1 | 2 - 3 | 643 | 77 | ||
| NM_001339751.1 | NP_001326017.1 | 2 - 3 | 322 | 77 | ||
| NM_115443.4 | NP_567027.2 | 2 - 3 | 328 | 77 | ||
| NM_202716.3 | NP_974445.2 | 3 - 4 | 541 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001339748.1, NM_001339749.1, NM_001339750.1 transcripts.
Set Membership: |
|

LAF3
Genomic Map