Assay Details
| Gene Symbol: LAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339748.1 | NP_001319763.1 | 3 - 4 | 1030 | 100 | |
| NM_001339751.1 | NP_001326017.1 | 4 - 5 | 508 | 100 | ||
| NM_115443.4 | NP_567027.2 | 4 - 5 | 514 | 100 | ||
| NM_202716.3 | NP_974445.2 | 5 - 6 | 727 | 100 | ||
More Information
Set Membership: |
|

LAF3
Genomic Map