Assay Details
| Gene Symbol: FER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115467.5 | NP_191168.1 | 5 - 6 | 655 | 89 | |
| GenBank mRNA | AJ312190.1 | - | 5 - 6 | 647 | 89 | |
| AK226273.1 | - | 5 - 6 | 653 | 89 | ||
| AY072221.1 | - | 5 - 6 | 652 | 89 | ||
| AY122951.1 | - | 5 - 6 | 604 | 89 | ||
More Information
Set Membership: |
|

FER3
Genomic Map