Assay Details
| Gene Symbol: ALPHA-SNAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115477.3 | NP_191178.1 | 2 - 3 | 539 | 70 | |
| GenBank mRNA | AF177989.1 | - | 2 - 3 | 200 | 70 | |
| BT024710.1 | - | 2 - 3 | 200 | 70 | ||
More Information
Set Membership: |
|

ALPHA-SNAP2
Genomic Map