Assay Details
| Gene Symbol: ALPHA-SNAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035795.1 | NP_001030872.1 | 3 - 4 | 444 | 100 | |
| NM_115477.3 | NP_191178.1 | 3 - 4 | 609 | 100 | ||
| GenBank mRNA | AF177989.1 | - | 3 - 4 | 270 | 100 | |
| BT024710.1 | - | 3 - 4 | 270 | 100 | ||
More Information
Set Membership: |
|

ALPHA-SNAP2
Genomic Map