Assay Details
| Gene Symbol: ALPHA-SNAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115503.1 | NP_191204.1 | 2 - 3 | 512 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALPHA-SNAP1| Gene Symbol: ALPHA-SNAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115503.1 | NP_191204.1 | 2 - 3 | 512 | 113 | |
Set Membership: |
|