Assay Details
| Gene Symbol: CIP111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115528.3 | NP_191228.1 | 4 - 5 | 1604 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CIP111| Gene Symbol: CIP111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115528.3 | NP_191228.1 | 4 - 5 | 1604 | 66 | |
Set Membership: |
|