Assay Details
| Gene Symbol: ACA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339853.1 | NP_001326564.1 | 3 - 4 | 2353 | 116 | |
| NM_115593.7 | NP_191292.1 | 3 - 4 | 2608 | 116 | ||
More Information
Set Membership: |
|

ACA11
Genomic Map