Assay Details
| Gene Symbol: ACA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339853.1 | NP_001326564.1 | 6 - 7 | 2984 | 125 | |
| NM_115593.7 | NP_191292.1 | 6 - 7 | 3239 | 125 | ||
More Information
Set Membership: |
|

ACA11
Genomic Map